Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRIM31 Partial ORF (NP_008959, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7099082
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to both the cytoplasm and the nucleus. Its function has not been identified. [provided by RefSeq]

Sequence: MASGQFVNKLQEEVICPICLDILQKPVTIDCGHNFCLKCITQIGETSCGFFKCPLCKTSVRKNAIRFNSLLRNLVEKIQALQASEVQSKRKEATCPRHQE

Specifications

Accession Number NP_008959
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 11074
Molecular Weight (g/mol) 36.74kDa
Name TRIM31 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MASGQFVNKLQEEVICPICLDILQKPVTIDCGHNFCLKCITQIGETSCGFFKCPLCKTSVRKNAIRFNSLLRNLVEKIQALQASEVQSKRKEATCPRHQE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias C6orf13/HCG1/HCGI/RNF
Common Name TRIM31
Gene Symbol TRIM31
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.