Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TRIM32 Antibody, R&D Systems™

Catalog No. MAB6515
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6515 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6515 Supplier R&D Systems Supplier No. MAB6515
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRIM32 Monoclonal specifically detects TRIM32 in Human samples. It is validated for Western Blot.

Specifications

Antigen TRIM32
Applications Western Blot
Classification Monoclonal
Clone 717801
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q13049
Gene Alias 72 kDa Tat-interacting protein, BBS11, BBS1172-KD, EC 6.3.2.-, HT2A, HT2Alimb girdle muscular dystrophy 2H (autosomal recessive), LGMD2H, TATIP, tripartite motif containing 32, Zinc finger protein HT2A, zinc-finger protein HT2A
Gene Symbols TRIM32
Host Species Mouse
Immunogen E. coli-derived recombinant human TRIM32 Arg105-Lys204 Accession # Q13049
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22954
Test Specificity Detects human TRIM32 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.