Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TRIM38 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | TRIM38 |
---|---|
Clone | 933011 |
Dilution | Immunohistochemistry 5-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9104SP
![]() |
R&D Systems
MAB9104SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9104100
![]() |
R&D Systems
MAB9104100 |
100 μg |
Each for $468.00
|
|
|||||
Description
TRIM38 Monoclonal specifically detects TRIM38 in Human samples. It is validated for Immunohistochemistry.Specifications
TRIM38 | |
Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MGC8946, RING finger protein 15RORETRo/SSA ribonucleoprotein homolog, RNF15, RoRet, tripartite motif containing 38, tripartite motif-containing 38, tripartite motif-containing protein 38, Zinc finger protein RoRet, zinc-finger protein RoRet | |
TRIM38 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
933011 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
O00635 | |
10475 | |
E.coli-derived recombinant human TRIM38 Leu190-Ile414 Accession # O00635 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TRIM38 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title