Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TRIM5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3684
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3684 100 μg
AF3684SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3684 Supplier R&D Systems Supplier No. AF3684
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

TRIM5 Polyclonal specifically detects TRIM5 in Human samples. It is validated for Western Blot.

Specifications

Antigen TRIM5
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9C035
Gene Alias EC 6.3.2, EC 6.3.2.-, RING finger protein 88, RNF88, RNF88tripartite motif protein TRIM, TRIM5alpha, tripartite motif containing 5, tripartite motif protein TRIM5, tripartite motif-containing 5, tripartite motif-containing protein 5
Gene Symbols TRIM5
Host Species Goat
Immunogen E. coli-derived recombinant human TRIM5 Ala2-Gln248 Accession # Q9C035
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85363
Test Specificity Detects human TRIM5 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse TRIM5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.