Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Tripeptidyl-Peptidase I/TPP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2237SP
This item is not returnable.
View return policy
Description
Tripeptidyl-Peptidase I/TPP1 Monoclonal specifically detects Tripeptidyl-Peptidase I/TPP1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Tripeptidyl-Peptidase I/TPP1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cell growth-inhibiting gene 1 protein, ceroid-lipofuscinosis, neuronal 2, late infantile (Jansky-Bielschowsky disease), CLN2, CLN2EC 3.4.14.9, growth-inhibiting protein 1, LINCL, LPIC, lysosomal pepstatin insensitive protease, Lysosomal pepstatin-insensitive protease, MGC21297, TPP1, TPP-1, TPP-I, Tripeptidyl aminopeptidase, tripeptidyl peptidase I, tripeptidyl-peptidase 1, Tripeptidyl-peptidase I, TripeptidylPeptidase I | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1200 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Immunoprecipitation | |
254008 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
O14773 | |
TPP1 | |
Mouse myeloma cell line NS0-derived recombinant human Tripeptidyl-Peptidase I/TPP1 Ser20-Pro563 Accession # O14773 | |
25 μg | |
Primary | |
Detects human Tripeptidyl-Peptidase I/TPP1 in direct ELISAs and Western blots. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction