Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TrkA Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FB1751RN100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB1751RN100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FB1751RN100 Supplier R&D Systems Supplier No. FAB1751RN100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TrkA Monoclonal specifically detects TrkA in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TrkA
Applications Flow Cytometry
Classification Monoclonal
Clone 165131R
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P04629
Gene Alias DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, NTRK1, NTRK-1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TrkA, Ala33-Glu407, Accession No. P04629
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4914.0
Test Specificity Detects human TrkA in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.