Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TrkA Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1752S100UG
This item is not returnable.
View return policy
Description
TrkA Monoclonal specifically detects TrkA in Human samples. It is validated for Flow Cytometry.Specifications
TrkA | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide | |
DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, NTRK1, NTRK-1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A | |
Mouse myeloma cell line NS0-derived recombinant human TrkA, Ala33-Glu407, Accession No. P04629 | |
Primary | |
Detects human TrkA in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
Flow Cytometry | |
994307 | |
Flow Cytometry 0.25-1 μg/106 cells | |
P04629 | |
Mouse | |
100 μg | |
4914 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction