Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TrkC Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $667.00
Specifications
Antigen | TrkC |
---|---|
Clone | 75213 |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Monoclonal |
Description
TrkC Monoclonal specifically detects TrkC in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
TrkC | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, EC 2.7.10.1, ETS related protein-neurotrophic receptor tyrosine kinase fusion protein, ETV6-NTRK3 fusion, gp145(trkC), GP145-TrkC, Neurotrophic tyrosine kinase receptor type 3, neurotrophic tyrosine kinase, receptor, type 3, NT 3 receptor, NT-3 growth factor receptor, NTRK3, Trk-C, TrkC tyrosine kinase, TRKCneurotrophin 3 receptor, tyrosine kinase receptor C | |
NTRK3 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
75213 | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
Q16288 | |
4916 | |
Mouse myeloma cell line NS0-derived recombinant human TrkC Cys32-Asp428 Accession # Q16288 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TrkC in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) TrkA, rhTrkB, or recombinant mouse TrkC is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title