Learn More
Human Troponin I Peptide 3 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Troponin I |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Hidden |
| Gene Symbols | TNNI3 |
| Host Species | Goat |
| Immunogen | Highly purified human cardiac troponin I |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.