Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Troponin I/TNNI3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | Troponin I Type 3 (cardiac) |
---|---|
Clone | 923952 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB85942SP
![]() |
R&D Systems
MAB85942SP |
25 μg |
Each for $200.50
|
|
|||||
MAB85942100
![]() |
R&D Systems
MAB85942100 |
100 μg |
Each for $514.50
|
|
|||||
Description
Troponin I Type 3 (cardiac) Monoclonal specifically detects Troponin I Type 3 (cardiac) in Human samples. It is validated for ELISA.Specifications
Troponin I Type 3 (cardiac) | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1, TNNC1troponin I, cardiac, TNNI3,, troponin I type 3 (cardiac), troponin I, cardiac muscle | |
E. coli-derived recombinant human Troponin I/TNNI3, Met1-Ser210 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human Troponin I/TNNI3 in direct ELISAs. |
923952 | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
7137 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title