Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Troponin T (Cardiac) Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
Supplier: R&D Systems MAB1874SP
This item is not returnable.
View return policy
Description
Cardiac Troponin T Monoclonal specifically detects Cardiac Troponin T in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
Cardiac Troponin T | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Cardiac Muscle Troponin T, Cardiomyopathy, Dilated 1D (Autosomal Dominant), Cardiomyopathy, Hypertrophic, CMD1D, CMH2, CMPD2, cTnT, LVNC6, RCM3, TNNT2, TnTC, troponin T type 2 (cardiac), troponin T, cardiac muscle, Troponin T2, Cardiac Type, Truncated Cardiac Troponin T | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7139 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Immunohistochemistry, Immunocytochemistry | |
200805 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
TNNT2 | |
Human cardiac Troponin T | |
25 μg | |
Primary | |
Detects purified human Troponin T (Cardiac) in direct ELISAs. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction