Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Troponin T (Cardiac) Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
Supplier: R&D Systems MAB1874
This item is not returnable.
View return policy
Description
Cardiac Troponin T Monoclonal specifically detects Cardiac Troponin T in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
Cardiac Troponin T | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Cardiac Muscle Troponin T, Cardiomyopathy, Dilated 1D (Autosomal Dominant), Cardiomyopathy, Hypertrophic, CMD1D, CMH2, CMPD2, cTnT, LVNC6, RCM3, TNNT2, TnTC, troponin T type 2 (cardiac), troponin T, cardiac muscle, Troponin T2, Cardiac Type, Truncated Cardiac Troponin T | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7139 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Immunohistochemistry, Immunocytochemistry | |
200805 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
TNNT2 | |
Human cardiac Troponin T | |
100 μg | |
Primary | |
Detects purified human Troponin T (Cardiac) in direct ELISAs. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction