Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TROY/TNFRSF19 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | TROY/TNFRSF19 |
---|---|
Clone | 933202 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB15481SP
![]() |
R&D Systems
MAB15481SP |
25 μg |
Each for $200.50
|
|
|||||
MAB15481100
![]() |
R&D Systems
MAB15481100 |
100 μg |
Each for $502.50
|
|
|||||
Description
TROY/TNFRSF19 Monoclonal specifically detects TROY/TNFRSF19 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
TROY/TNFRSF19 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
TAJ, TAJ-alpha, TAJToxicity and JNK inducer, TNFRSF19, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
Mouse myeloma cell line NS0-derived recombinant human TROY/TNFRSF19 Glu30-Leu170 Accession # Q9NS68 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TROY/TNFRSF19 in direct ELISAs. Stains human TROY/TNFRSF19 transfectants but not irrelevant transfectants in flow cytometry. |
933202 | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
Q9NS68 | |
55504 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title