Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Trypsin 1/PRSS1 ELISA Kit

Catalog No. EH468RBX5
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests

ELISA

Human Trypsin 1/PRSS1 quantitates human Trypsin 1/PRSS1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Trypsin 1/PRSS1.

Trypsin is a 24 kDa enzyme of the serine proteinase family. It is produced in the pancreas as an inactive precursor, trypsinogen, but the active enzyme is located in the gastrointestinal tract where it degrades proteins to large peptides. Trypsin prefers lysine and arginine residues. One of the substrates for trypsin is chymotrypsinogen which is cleaved to produce active chymotrypsin. High levels of immunoreactive trypsin in the bloodstream can indicate pancreatic malfunction and this indicator is used to screene for and diagnose Cystic Fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Alpha-trypsin chain 1; Alpha-trypsin chain 2; anionic trypsin I; Anionic trypsin-1; beta-trypsin; Cationic trypsinogen; digestive zymogen; MGC120175; MGC149362; nonfunctional trypsin 1; pancreatic trypsin 1; pretrypsinogen I; Protease Serine 1; protease, serine 1; protease, serine 1 (trypsin 1); protease, serine, 1; protease, serine, 1 (trypsin 1); protease, serine, 2; PRSS1; PTRYI; RATPTRYI; Serine protease 1; TCR V beta 4.1; TRP1; TRY1; Try-1; TRY4; Trygn16; TRYP1; TRYP8; trypsin 1; Trypsin I; Trypsin-1; trypsinogen; trypsinogen 1; trypsinogen 16; trypsinogen A
Assay Range 0.29 to 200 ng/mL
Assay Sensitivity 0.29 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5644
Gene Symbol PRSS1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Trypsin
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.