Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Trypsin Pan Specific (PRSS1/2/3) Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF3586
This item is not returnable.
View return policy
Description
Trypsin Pan Specific/PRSS1/2/3 Polyclonal specifically detects Trypsin Pan Specific/PRSS1/2/3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Neutralization.Specifications
Trypsin Pan Specific/PRSS1/2/3 | |
Polyclonal | |
Unconjugated | |
P07478 | |
Sheep | |
Affinity Purified | |
RUO | |
5645 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Immunoprecipitation, Neutralization | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 12.5 ug/500 ug cell lysate, Neutralization 0.3 ug/mL | |
Mesotrypsin, MTG, PRSS1, PRSS2, PRSS3, PRSS4, PTRYI, Tesp4, TRY2, TRY4, TRY8, Trygn16, TRYP2, Trypsin 4, Trypsinogen 1, Trypsinogen 15, Trypsinogen 20 | |
Mouse myeloma cell line NS0-derived recombinant human Trypsin 2 Ala16-Ser247 Accession # P07478 | |
100 μg | |
Primary | |
Detects human Trypsin Pan Specific (PRSS1/2/3) in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction