Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Tryptase alpha/TPS1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2370
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2370 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2370 Supplier R&D Systems Supplier No. BAF2370
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Tryptase alpha/TPS1 Polyclonal specifically detects Tryptase alpha/TPS1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Tryptase alpha/TPS1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. AAA86934
Gene Alias alpha-Tryptase, TPS1, TPS2, TPSAB1, TPSB1, Tryptase 1, tryptase alpha/beta 1
Gene Symbols TPSAB1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Tryptase alpha/TPS1 Ile31-Pro275 Accession # AAA86934
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7177
Test Specificity Detects human Tryptase alpha/TPS1 in Western blots. In Western blots, approximately 60% cross-reactivity with recombinant human (rh) TPSB2 is observed and approximately 10% cross-reactivity with rhTPSG1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.