Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Tryptase beta-2/TPSB2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3796
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3796 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3796 Supplier R&D Systems Supplier No. MAB3796
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tryptase beta-2/TPSB2 Monoclonal specifically detects Tryptase beta-2/TPSB2 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Tryptase beta-2/TPSB2
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 349406
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAD13876
Gene Alias EC 3.4.21, EC 3.4.21.59, mast cell tryptase beta II, mast cell tryptase beta III, TPS2, TPS2tryptase beta-2, TPSB2, tryptase beta 2, tryptase beta 2 (gene/pseudogene), tryptase beta II, tryptase beta III, Tryptase beta2, Tryptase II, tryptase III, tryptase-2, tryptaseB, tryptaseC, Tryptase-C
Gene Symbols TPSB2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Tryptase beta-2/TPSB2 Ile31-Pro275 Accession # AAD13876
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64499
Test Specificity Detects human Tryptase beta-2/TPSB2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 50-100% cross-reactivity with recombinant human (rh) TPSAB1 and recombinant mouse (rm) Mcpt6 is observed, 20-100% cross-reactivity with rmMcpt7 is observed, and no cross-reactivity with rhTPSG1, rmTPS5, rmMcpt1, or rmMcpt11 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.