Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Tryptase epsilon/BSSP-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5047SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5047SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5047SP Supplier R&D Systems Supplier No. MAB5047SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tryptase epsilon/BSSP-4/PRSS22 Monoclonal specifically detects Tryptase epsilon/BSSP-4/PRSS22 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Tryptase epsilon/BSSP-4/PRSS22
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 725445
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9GZN4
Gene Alias BSP-2, BSSP4, BSSP-4, hBSSP-4, MGC9599, prosemin, protease, serine S1 family member 22, protease, serine, 22, PRSS22, serine protease 22, serine protease 26, SP001LA, tryptase epsilon, UNQ302/PRO343
Gene Symbols PRSS22
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Tryptase epsilon/BSSP-4 Ala33-Ser317 Accession # Q9GZN4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64063
Test Specificity Detects human Tryptase epsilon/BSSP-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Tryptase epsilon/BSSP-4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.