Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Tryptase gamma-1/TPSG1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1667SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1667SP 25 μg
MAB1667 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1667SP Supplier R&D Systems Supplier No. MAB1667SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Tryptase gamma-1/TPSG1 Monoclonal specifically detects Tryptase gamma-1/TPSG1 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Tryptase gamma-1/TPSG1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 279005
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9NRR2
Gene Alias EC 3.4.21, EC 3.4.21.-, gamma I, gamma II, lung tryptase, mast cell protease II, mast cell tryptase, PRSS31skin tryptase, Serine protease 31, TMTpituitary tryptase, TPSG1, Transmembrane tryptase, trpA, tryptase gamma, tryptase gamma 1, tryptase gamma I, tryptase gamma II, Tryptase gamma1
Gene Symbols TPSG1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Tryptase gamma-1/TPSG1 Arg20-Arg281 Accession # Q9NRR2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25823
Test Specificity Detects human Tryptase gamma-1/TPSG1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) MCPT-1, recombinant human (rh) Tryptase alpha , rmTryptase beta-1, rhTryptase beta-2, or rmTryptase-5 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.