Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TSC1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC4379V100UG
This item is not returnable.
View return policy
Description
TSC1 Monoclonal specifically detects TSC1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
TSC1 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q92574 | |
TSC1 | |
E. coli-derived recombinant human TSC1 Asp156-Thr300 Accession # Q92574 | |
RUO | |
7248 | |
Human | |
IgG2b |
Flow Cytometry | |
488915 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Hamartin, KIAA0243MGC86987, LAMhamartin, TSC, tuberous sclerosis 1, Tuberous sclerosis 1 protein, tumor suppressor | |
Mouse | |
100 μg | |
Primary | |
Detects human TSC1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) TSC2 (aa 550-850), rhTSC2 (aa 1506-1748), or recombinant mouse TSC22 (aa 1-143) is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction