Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TSC1 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC4379N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4379N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC4379N100 Supplier R&D Systems Supplier No. IC4379N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TSC1 Monoclonal specifically detects TSC1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen TSC1
Applications Flow Cytometry
Classification Monoclonal
Clone 488915
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q92574
Gene Alias Hamartin, KIAA0243MGC86987, LAMhamartin, TSC, tuberous sclerosis 1, Tuberous sclerosis 1 protein, tumor suppressor
Gene Symbols TSC1
Host Species Mouse
Immunogen E. coli-derived recombinant human TSC1 Asp156-Thr300 Accession # Q92574
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7248
Test Specificity Detects human TSC1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) TSC2 (aa 550-850), rhTSC2 (aa 1506-1748), or recombinant mouse TSC22 (aa 1-143) is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.