Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TSC2 Antibody, R&D Systems™

Catalog No. AF4040
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4040 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4040 Supplier R&D Systems Supplier No. AF4040
This item is not returnable. View return policy

Sheep Polyclonal Antibody

TSC2 Polyclonal specifically detects TSC2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TSC2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P49815
Gene Alias LAM, LAMFLJ43106, TSC4, Tuberin, tuberous sclerosis 2, Tuberous sclerosis 2 protein
Gene Symbols TSC2
Host Species Sheep
Immunogen E. coli-derived recombinant human TSC2 His1506-Lys1748 Accession # P49815
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7249
Test Specificity Detects human TSC2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human TSC1 and recombinant mouse TSC22 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.