Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TSEN34 Partial ORF (NP_076980, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7101969
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

tRNA splicing is a fundamental process required for cell growth and division. SEN34 is a subunit of the tRNA splicing endonuclease, which catalyzes the removal of introns, the first step in tRNA splicing (Paushkin et al., 2004 [PubMed 15109492]).[supplied by OMIM]

Sequence: MLVVEVANGRSLVWGAEAVQALRERLGVGGRTVGALPRGPRQNSRLGLPLLLMPEEARLLAEIGAVTLVSAPRPDSRHHSLALTSFKRQQEESFQEQSA

Specifications

Accession Number NP_076980
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 79042
Molecular Weight (g/mol) 36.63kDa
Name TSEN34 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MLVVEVANGRSLVWGAEAVQALRERLGVGGRTVGALPRGPRQNSRLGLPLLLMPEEARLLAEIGAVTLVSAPRPDSRHHSLALTSFKRQQEESFQEQSA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias LENG5/PCH2C/SEN34/SEN34L
Common Name TSEN34
Gene Symbol TSEN34
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.