Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TSG101 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. RD10000180
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10000180 100 μg
MAB114381SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. RD10000180 Supplier R&D Systems Supplier No. MAB114381100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TSG101 Monoclonal antibody specifically detects TSG101 in Human samples. It is validated for Western Blot

Specifications

Antigen TSG101
Applications Western Blot
Classification Monoclonal
Clone 1065916
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q99816
Gene Alias ESCRT-I complex subunit TSG101, TSG10, tumor susceptibility gene 10, tumor susceptibility gene 101, tumor susceptibility gene 101 protein, tumor susceptibility protein, VPS23
Host Species Mouse
Immunogen TSG101 containing synthetic peptide, Accession # Q99816
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7251
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.