Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TSLP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1398
This item is not returnable.
View return policy
Description
TSLP Monoclonal specifically detects TSLP in Human samples. It is validated for Western Blot.Specifications
TSLP | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
Q969D9 | |
TSLP | |
E. coli-derived recombinant human TSLP Tyr29-Gln159 Accession # Q969D9 | |
100 μg | |
Primary | |
Detects human TSLP in direct ELISAs and Western blots. Does not cross-react with recombinant mouse (rm) IL-7 or rmTSLP. | |
Human | |
Purified |
Western Blot | |
258136 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
thymic stromal lymphopoietin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
85480 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction