Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TSLP Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.28 - $567.90
Specifications
| Antigen | TSLP |
|---|---|
| Clone | 933829 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB13982-SP
![]() |
R&D Systems
MAB13982SP |
25 μg |
Each for $188.28
|
|
|||||
MAB13982-100
![]() |
R&D Systems
MAB13982100 |
100 μg |
Each for $567.90
|
|
|||||
Description
TSLP Monoclonal specifically detects TSLP in Human samples. It is validated for ELISA.Specifications
| TSLP | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Q969D9 | |
| 85480 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 933829 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| thymic stromal lymphopoietin | |
| E. coli-derived recombinant human TSLP Tyr29-Gln159 Accession # Q969D9 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human TSLP in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title