Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TSLPR Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $410.00
Specifications
Antigen | TSLPR/CRLF2 |
---|---|
Clone | 147036R |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB981RSP
![]() |
R&D Systems
MAB981RSP |
25 μg |
Each for $241.00
|
|
|||||
MAB981R100
![]() |
R&D Systems
MAB981R100 |
100 μg |
Each for $410.00
|
|
|||||
Description
TSLPR/CRLF2 Monoclonal specifically detects TSLPR/CRLF2 in Human samples. It is validated for Western Blot.Specifications
TSLPR/CRLF2 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
CRL2, CRL2cytokine receptor CRL2 precusor, CRLF2, CRLF2Y, CRLM-2, Cytokine receptor-like 2, cytokine receptor-like factor 2, ILXR, IL-XR, P2RY8/CRLF2 fusion, PCOR1, Thymic stromal lymphopoietin protein receptor, thymic stromal lymphopoietin receptor, thymic stromal-derived lymphopoietin receptor, TSLP R, TSLP receptor, TSLPR | |
Mouse myeloma cell line NS0-derived recombinant human TSLP R, Gly25-Lys231, Accession # Q9HC73 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
147036R | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9HC73 | |
64109 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title