Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TSP50 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2430
This item is not returnable.
View return policy
Description
TSP50 Polyclonal specifically detects TSP50 in Human samples. It is validated for Western Blot.Specifications
TSP50 | |
Polyclonal | |
Biotin | |
Q9UI38 | |
PRSS50 | |
Mouse myeloma cell line NS0-derived recombinant human TSP50 Ala40-Ala363 Accession # Q9UI38 | |
50 μg | |
Primary | |
Detects human TSP50 in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant mouse TSP50 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Cancer/testis antigen 20, CT20probable threonine protease PRSS50, EC 3.4.25.-, protease, serine, 50, Serine protease 50, testes-specific protease 50, Testis-specific protease-like protein 50, TSP50testes specific protease 50 | |
Goat | |
Affinity Purified | |
RUO | |
29122 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction