Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TSPAN12 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8910R025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8910R025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8910R025 Supplier R&D Systems Supplier No. FAB8910R025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TSPAN12 Monoclonal specifically detects TSPAN12 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TSPAN12
Applications Flow Cytometry
Classification Monoclonal
Clone 921938
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. O95859
Gene Alias EVR5, Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, NET2, NET-2, ten-eleven translocation-1, tetraspanin-12, TM4SF12
Gene Symbols TSPAN12
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human TSPAN12 Met1-Leu305 Accession # O95859
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23554
Test Specificity Stains human TSPAN12 transfectants but not irrelevant transfectants in flow cytometry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.