Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TSPAN12 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8910S100UG
This item is not returnable.
View return policy
Description
TSPAN12 Monoclonal specifically detects TSPAN12 in Human samples. It is validated for Flow Cytometry.Specifications
TSPAN12 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Mouse | |
100 μg | |
Primary | |
Detects human TSPAN12 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
921938 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
EVR5, Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, NET2, NET-2, ten-eleven translocation-1, tetraspanin-12, TM4SF12 | |
NS0 mouse myeloma cell line transfected with human TSPAN12 Met1-Leu305 | |
RUO | |
23554 | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction