Learn More
Human TSPAN2 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | TSPAN2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 822509 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 405 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | O60636 |
| Gene Alias | FLJ12082, NET3, new EST tetraspan 3, tetraspan 2, Tetraspan NET-3, tetraspan TM4SF, tetraspanin 2, tetraspanin 2 isoform, tetraspanin-2, TSN2, Tspan-2 |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.