Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TSPAN8 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB4734T100UG
This item is not returnable.
View return policy
Description
TSPAN8/TM4SF3 Monoclonal specifically detects TSPAN8/TM4SF3 in Human samples. It is validated for Flow Cytometry.Specifications
TSPAN8/TM4SF3 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAH05246 | |
TSPAN8 | |
NS0 mouse myeloma cell line transfected with human TSPAN8 Met1-Lys237 (Ile35Val) Accession # AAH05246 | |
RUO | |
7103 | |
Human | |
IgG2b |
Flow Cytometry | |
458811 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CO-029, tetraspanin 8, TM4SF3, TM4SF3tspan-8, Transmembrane 4 superfamily member 3tetraspanin-8, Tspan-8, Tumor-associated antigen CO-029 | |
Rat | |
100 μg | |
Primary | |
Detects human TSPAN8 in Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction