Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TTF-1/NKX2-1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.16 - $544.12
Specifications
| Antigen | TTF-1 / NKX2-1 |
|---|---|
| Clone | 2054B |
| Dilution | Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9458SP
![]() |
R&D Systems
MAB9458SP |
25 μg |
Each for $200.16
|
|
|||||
MAB9458100
![]() |
R&D Systems
MAB9458100 |
100 μg |
Each for $544.12
|
|
|||||
Description
TTF-1 / NKX2-1 Monoclonal specifically detects TTF-1 / NKX2-1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| TTF-1 / NKX2-1 | |
| Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P43699 | |
| 7080 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 2054B | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BCH, benign chorea, BHC, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2.1, NKX2-1, NKX2A, NKX2ANKX2.1, T/EBP, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2 | |
| E. coli-derived recombinant human TTF-1/NKX2-1 Ala40-Pro111 Accession # P43699 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human NKX2.1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title