Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TTF-1/NKX2-1 Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

$214.00 - $573.00

Specifications

Antigen TTF-1 / NKX2-1
Clone 2054E
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 3-25 ug/mL
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB94581SP
GREENER_CHOICE
View Documents
R&D Systems
MAB94581SP
25 μg
Each for $214.00
Only null left
Add to Cart
 
MAB94581100
GREENER_CHOICE
View Documents
R&D Systems
MAB94581100
100 μg
Each for $573.00
Only null left
Add to Cart
 
Description

Description

TTF-1 / NKX2-1 Monoclonal specifically detects TTF-1 / NKX2-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications

Specifications

TTF-1 / NKX2-1
Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 3-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
BCH, benign chorea, BHC, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2.1, NKX2-1, NKX2A, NKX2ANKX2.1, T/EBP, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2
E. coli-derived recombinant human TTF-1/NKX2-1 Ala40-Pro111 Accession # P43699
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human NKX2.1 in direct ELISAs.
2054E
Western Blot, Immunohistochemistry, Immunocytochemistry
Unconjugated
Rabbit
Human
P43699
7080
IgG
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only (RUO)

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.