Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Tumstatin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7546SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7546SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7546SP Supplier R&D Systems Supplier No. MAB7546SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tumstatin/COL4A3 Monoclonal specifically detects Tumstatin/COL4A3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Tumstatin/COL4A3
Applications Western Blot
Classification Monoclonal
Clone 762102
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias COL4A3, collagen alpha-3(IV) chain, collagen IV, alpha-3 polypeptide, collagen, type IV, alpha 3 (Goodpasture antigen), Goodpasture antigen, tumstatin
Gene Symbols COL4A3
Host Species Mouse
Immunogen E. coli-derived recombinant human Tumstatin
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1285
Test Specificity Detects human Tumstatin in direct ELISAs. In direct ELISAs, less than 25% cross-reactivity with recombinant human Collagen IV alpha 1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.