Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TWEAK R/TNFRSF12 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF1199SP
This item is not returnable.
View return policy
Description
TWEAK R/TNFRSF12 Polyclonal specifically detects TWEAK R/TNFRSF12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
TWEAK R/TNFRSF12 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, Fn14, FN14tumor necrosis factor receptor superfamily member 12A, TNFRSF12, TNFRSF12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14 | |
Goat | |
Affinity Purified | |
RUO | |
51330 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.3-1.2 ug/mL | |
Q9NP84 | |
TNFRSF12A | |
recombinant human TWEAK R/TNFRSF12 Glu28-Trp79 Accession # Q9NP84 | |
25 μg | |
Primary | |
Detects human TWEAK R/TNFRSF12 in direct ELISAs and Western blots. In direct ELISAs, approximately 80% cross-reactivity with recombinant mouse TWEAK R/TNFRSF12 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction