Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TWEAK R/TNFRSF12 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF1199
This item is not returnable.
View return policy
Description
TWEAK R/TNFRSF12 Polyclonal specifically detects TWEAK R/TNFRSF12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
| TWEAK R/TNFRSF12 | |
| Polyclonal | |
| Unconjugated | |
| Q9NP84 | |
| TNFRSF12A | |
| recombinant human TWEAK R/TNFRSF12 Glu28-Trp79 Accession # Q9NP84 | |
| 100 μg | |
| Primary | |
| Detects human TWEAK R/TNFRSF12 in direct ELISAs and Western blots. In direct ELISAs, approximately 80% cross-reactivity with recombinant mouse TWEAK R/TNFRSF12 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.3-1.2 ug/mL | |
| CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, Fn14, FN14tumor necrosis factor receptor superfamily member 12A, TNFRSF12, TNFRSF12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 51330 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction