Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TWEAK/TNFSF12 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB1090100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1090100 100 μg
MAB1090SP 25 μg
MAB1090500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1090100 Supplier R&D Systems Supplier No. MAB1090100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TWEAK/TNFSF12 Monoclonal specifically detects TWEAK/TNFSF12 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen TWEAK/TNFSF12
Applications ELISA Capture (Matched Antibody Pair)
Classification Monoclonal
Clone 173830
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O43508
Gene Alias APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, TNFSF12, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12
Gene Symbols TNFSF12
Host Species Mouse
Immunogen E. coli-derived recombinant human TWEAK/TNFSF12 Arg93-His249 Accession # O43508
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8742
Test Specificity Detects human TWEAK/TNFSF12 in ELISAs. In sandwich immunoassays, approximately 15% reactivity with recombinant mouse TWEAK is observed and no cross-reactivity with recombinant human (rh) BAFF, rhEDA-A2, rhLIGHT, rhFas Ligand, rhGITR Ligand, rhLT alpha 1 beta 2, rhLT alpha 2 beta 1, rhOX40 Ligand, rhTNF-alpha , rhTRANCE, rhTRAIL, rh4-1BB, or rhAPRIL is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Form Supernatant
Isotype IgG1
Product Line R&D Systems
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.