Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Twist-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6230RSP
This item is not returnable.
View return policy
Description
Twist-1 Monoclonal specifically detects Twist-1 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
Twist-1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
acrocephalosyndactyly 3, ACS3, B-HLH DNA binding protein, BHLHA38, bHLHa38BPES3, BPES3, Class A basic helix-loop-helix protein 38, CRS1, H-twist, H-twistblepharophimosis, epicanthus inversus and ptosis 3, SCSBPES2, TWIST, twist homolog 1 (Drosophila), TWIST homolog of drosophila, Twist1, twist-related protein 1 | |
E.coli-derived recombinant human Twist-1 Met1-His202 Accession # Q15672 | |
25 μg | |
7291 | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
CyTOF, Flow Cytometry, Western Blot | |
927403R | |
Western Blot 2μg/mL, Intracellular Staining by Flow Cytometry 0.25μg/106 cells, CyTOF-ready | |
Q15672 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human Twist-1 in direct ELISAs. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction