Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Twist-1 Fluorescein-conjugated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems IC6230F
This item is not returnable.
View return policy
Description
Twist-1 Polyclonal specifically detects Twist-1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Twist-1 | |
Polyclonal | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
acrocephalosyndactyly 3, ACS3, B-HLH DNA binding protein, BHLHA38, bHLHa38BPES3, BPES3, Class A basic helix-loop-helix protein 38, CRS1, H-twist, H-twistblepharophimosis, epicanthus inversus and ptosis 3, SCSBPES2, TWIST, twist homolog 1 (Drosophila), TWIST homolog of drosophila, Twist1, twist-related protein 1 | |
Sheep | |
Affinity Purified | |
RUO | |
7291 | |
Human | |
IgG |
Flow Cytometry | |
Fluorescein | |
Q15672 | |
TWIST1 | |
E. coli-derived recombinant human Twist-1 Met1-His202 Accession # Q15672 | |
100 Tests | |
Primary | |
Detects human Twist-1 in direct ELISAs. In direct ELISAs, less than 16% cross-reactivity with recombinant human Twist-2 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction