Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Twist-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6249
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6249 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6249 Supplier R&D Systems Supplier No. AF6249
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Twist-2 Polyclonal specifically detects Twist-2 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen Twist-2
Applications Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q8WVJ9
Gene Alias BHLHA39, bHLHa39twist-related bHLH protein Dermo1, Class A basic helix-loop-helix protein 39, Dermis-expressed protein 1, Dermo-1, DERMO1, DERMO1MGC117334, twist homolog 2 (Drosophila), Twist2, twist-related protein 2
Gene Symbols TWIST2
Host Species Sheep
Immunogen E. coli-derived recombinant human Twist-2 Met1-Ser159 Accession # Q8WVJ9
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117581
Test Specificity Detects human Twist-2 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant human Twist-1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.