Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Tyk2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3427
This item is not returnable.
View return policy
Description
Tyk2 Monoclonal specifically detects Tyk2 in Human samples. It is validated for Western Blot.Specifications
Tyk2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P29597 | |
TYK2 | |
E. coli-derived recombinant human Tyk2 Pro2-Leu51 Accession # P29597 | |
100 μg | |
Primary | |
Detects human Tyk2 in Western blots. | |
Human | |
Purified |
Western Blot | |
307214 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, EC 2.7.10.2, JTK1, non-receptor tyrosine-protein kinase TYK2, tyrosine kinase 2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7297 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction