Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Tyro3/Dtk Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB859R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB859R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB859R100 Supplier R&D Systems Supplier No. FAB859R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dtk/TYRO3 Monoclonal specifically detects Dtk/TYRO3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Dtk/TYRO3
Applications Flow Cytometry
Classification Monoclonal
Clone 96201
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1μg/106 cells
Formulation Supplied 0.2mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q06418
Gene Alias Brt, BYK, Dtk, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Tyro3/Dtk Ala41-Ser428 (predicted) Accession # Q06418
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7301
Test Specificity Detects human Tyro3/Dtk in ELISAs and Western blots. In ELISAs and Western blots, no cross-reactivity or interferance with recombinant mouse Tyro3/Dtk, recombinant human (rh) Axl, or rhMer is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.