Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Tyro3/Dtk Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB859SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB859SP 25 μg
MAB859100 100 μg
MAB859500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB859SP Supplier R&D Systems Supplier No. MAB859SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 8 publications

Dtk/TYRO3 Monoclonal specifically detects Dtk/TYRO3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Knockout Validated, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Dtk/TYRO3
Applications Flow Cytometry, Immunohistochemistry, Immunoassay, ELISA
Classification Monoclonal
Clone 96201
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Knockout Validated 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q06418
Gene Alias Brt, BYK, Dtk, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
Gene Symbols TYRO3
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Tyro3/Dtk Ala41-Ser428 (predicted) Accession # Q06418
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7301
Test Specificity Detects human Tyro3/Dtk in ELISAs and Western blots. In ELISAs and Western blots, no cross-reactivity or interferance with recombinant mouse Tyro3/Dtk, recombinant human (rh) Axl, or rhMer is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.