Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UBE2J2 Full-length ORF (NP_477515.2, 1 a.a. - 259 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89966406 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-406 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-966-406 Supplier Abnova™ Supplier No. H00118424P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is located in the membrane of the endoplasmic reticulum. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined. [provided by RefSeq]

Sequence: MSSTSSKRAPTTATQRLKQDYLRIKKDPVPYICAEPLPSNILEWHYVVRGPEMTPYEGGYYHGKLIFPREFPFKPPSIYMITPNGRFKCNTRLCLSITDFHPDTWNPAWSVSTILTGLLSFMVEKGPTLGSIETSDFTKRQLAVQSLAFNLKDKVFCELFPEVVEEIKQKQKAQDELSSRPQTLPLPDVVPDGETHLVQNGIQLLNGHAPGAVPNLAGLQQANRHHGLLGGALANLFVIVGFAAFAYTVKYVLRSIAQE

Specifications

Accession Number NP_477515.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 118424
Molecular Weight (g/mol) 55.3kDa
Name UBE2J2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MSSTSSKRAPTTATQRLKQDYLRIKKDPVPYICAEPLPSNILEWHYVVRGPEMTPYEGGYYHGKLIFPREFPFKPPSIYMITPNGRFKCNTRLCLSITDFHPDTWNPAWSVSTILTGLLSFMVEKGPTLGSIETSDFTKRQLAVQSLAFNLKDKVFCELFPEVVEEIKQKQKAQDELSSRPQTLPLPDVVPDGETHLVQNGIQLLNGHAPGAVPNLAGLQQANRHHGLLGGALANLFVIVGFAAFAYTVKYVLRSIAQE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias NCUBE2/PRO2121
Common Name UBE2J2
Gene Symbol UBE2J2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.