Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ubiquitin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$241.00 - $591.44
Specifications
| Antigen | Ubiquitin |
|---|---|
| Clone | 1002A |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Ubiquitin Monoclonal specifically detects Ubiquitin in Human samples. It is validated for Western Blot.Specifications
| Ubiquitin | |
| Western Blot 0.1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with Sodium Azide | |
| RPS27A, UBA52, UBB, UBB ubiquitin B, UBC | |
| UBB | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 1002A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P0CG47 | |
| 6233 | |
| HEK293 human embryonic kidney cell line transfected with human Ubiquitin Accession # P0CG47 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Ubiquitin in direct ELISAs |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title