Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human UCH-L1/PGP9.5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6007
This item is not returnable.
View return policy
Description
UCH-L1/PGP9.5 Monoclonal specifically detects UCH-L1/PGP9.5 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Knockout Validated.Specifications
UCH-L1/PGP9.5 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL | |
P09936 | |
UCHL1 | |
E. coli-derived recombinant human UCH-L1/PGP9.5 Gln2-Ala223 Accession # P09936 | |
100 μg | |
Primary | |
Detects human UCH-L1/PGP9.5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human UCH-L3 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
671108 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7345 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction