Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ULBP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 42 publications
$214.00 - $2020.00
Specifications
Antigen | ULBP-1 |
---|---|
Clone | 170818 |
Concentration | LYOPH |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready |
Applications | Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1380SP
![]() |
R&D Systems
MAB1380SP |
25 μg |
Each for $214.00
|
|
|||||
MAB1380100
![]() |
R&D Systems
MAB1380100 |
100 μg |
Each for $549.50
|
|
|||||
MAB1380500
![]() |
R&D Systems
MAB1380500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
ULBP-1 Monoclonal specifically detects ULBP-1 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
ULBP-1 | |
LYOPH | |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
ALCAN-beta, MULT-1, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL, NKG2DL1, RAET1I, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1, ULBP1 | |
ULBP1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
170818 | |
Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
80329 | |
BaF3 mouse pro-B cell line transfected with human ULBP-1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Stains human ULBP-1 cell transfectants. It does not stain cells transfected with ULBP-2 or ULBP-3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title