Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ULBP-2/5/6 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1298T100UG
This item is not returnable.
View return policy
Description
ULBP-2/5/6 Monoclonal specifically detects ULBP-2/5/6 in Human samples. It is validated for Flow Cytometry.Specifications
ULBP-2/5/6 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9BZM5 | |
Mouse | |
100 μg | |
Primary | |
Detects human ULBP-2 and human RAET1L/ULBP-6 in direct ELISA and stains cells transfected with human ULBP-2, human ULBP-5 or human RAET1L/ULBP-6 in flow cytometry. In direct ELISA, 11% cross-reactivity with recombinant human ULBP5 is observed. It does not stain cells transfected with human ULBP-1 or human ULBP-3 in flow cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
165903 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
ALCAN-alpha, N2DL2, NKG2DL2, RAET1G, RAET1H, RAET1L, UL16 binding protein 2, ULBP2, ULBP5, ULBP6 | |
BaF3 mouse pro-B cell line transfected with human ULBP-2 Accession # Q9BZM5 | |
RUO | |
80328 | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction