Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ULBP-2/5/6 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 43 publications
$227.50 - $2070.00
Specifications
Antigen | ULBP-2/5/6 |
---|---|
Clone | 165903 |
Concentration | LYOPH |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 1 ug/mL, CyTOF-ready |
Applications | Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1298SP
![]() |
R&D Systems
MAB1298SP |
25 μg |
Each for $227.50
|
|
|||||
MAB1298100
![]() |
R&D Systems
MAB1298100 |
100 μg |
Each for $573.00
|
|
|||||
MAB1298500
![]() |
R&D Systems
MAB1298500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
ULBP-2/5/6 Monoclonal specifically detects ULBP-2/5/6 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
ULBP-2/5/6 | |
LYOPH | |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
Q9BZM5 | |
80328 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
165903 | |
Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 1 ug/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ALCAN-alpha, N2DL2, NKG2DL2, RAET1G, RAET1H, RAET1L, UL16 binding protein 2, ULBP2, ULBP5, ULBP6 | |
BaF3 mouse pro-B cell line transfected with human ULBP-2 Accession # Q9BZM5 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ULBP-2 and human RAET1L/ULBP-6 in direct ELISA and stains cells transfected with human ULBP-2, human ULBP-5 or human RAET1L/ULBP-6 in flow cytometry. In direct ELISA, 11% cross-reactivity with recombinant human ULBP5 is observed. It does not stain cells transfected with human ULBP-1 or human ULBP-3 in flow cytometry. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title